siRNA Details
| Gene Symbol: Arel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106744.1 | NP_001100214.1 | 15 | 2119 | ||
| XM_006240333.2 | XP_006240395.1 | 16 | 3105 | |||
| XM_006240335.2 | XP_006240397.1 | 16 | 3075 | |||
| XM_008764754.1 | XP_008762976.1 | 15 | 2380 | |||
| XM_008764755.1 | XP_008762977.1 | 17 | 3212 | |||
| GenBank mRNA | FQ226603.1 | - | 7 | 698 | ||

Arel1
Genomic Map