siRNA Details
| Gene Symbol: Dlst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006981.2 | NP_001006982.2 | 9, 10 | 718 | ||
| XM_006240306.1 | XP_006240368.1 | 7, 8 | 750 | |||
| XM_008764756.1 | XP_008762978.1 | 8, 9 | 649 | |||
| GenBank mRNA | AB504752.1 | - | 2, 3 | 123 | ||
| BC083858.1 | - | 9, 10 | 704 | 7125734 | ||
| CK355938.1 | - | 9, 10 | 721 | |||
| D90401.1 | - | 9, 10 | 669 | |||
| FQ217447.1 | - | 8, 9 | 643 | |||
| FQ217539.1 | - | 8, 9 | 643 | |||

Dlst
Genomic Map