siRNA Details
| Gene Symbol: RGD1310769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106749.1 | NP_001100219.1 | 4, 5 | 813 | ||
| XM_006240367.2 | XP_006240429.1 | 3, 4 | 622 | |||
| XM_006240368.1 | XP_006240430.1 | 3, 4 | 341 | |||
| GenBank mRNA | FQ212891.1 | - | 3, 4 | 301 | ||

RGD1310769
Genomic Map