siRNA Details
| Gene Symbol: Ift43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134525.1 | NP_001127997.1 | 7 | 459 | ||
| XM_006240370.2 | XP_006240432.1 | 8 | 529 | |||
| XM_006240372.2 | XP_006240434.1 | 7 | 445 | |||
| GenBank mRNA | FQ222650.1 | - | 3 | 180 | ||

Ift43
Genomic Map