siRNA Details
| Gene Symbol: Ift43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134525.1 | NP_001127997.1 | 6, 7 | 422 | ||
| XM_006240370.2 | XP_006240432.1 | 7, 8 | 492 | |||
| XM_006240372.2 | XP_006240434.1 | 6, 7 | 408 | |||
| GenBank mRNA | FQ222650.1 | - | 2, 3 | 143 | ||

Ift43
Genomic Map