siRNA Details
| Gene Symbol: Esrrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008516.2 | NP_001008516.2 | 3, 4 | 634 | ||
| XM_006240374.2 | XP_006240436.1 | 4, 5 | 1096 | |||
| XM_006240375.2 | XP_006240437.1 | 4, 5 | 1097 | |||
| XM_008764759.1 | XP_008762981.1 | 4, 5 | 2332 | |||
| GenBank mRNA | AY383731.1 | - | 3, 4 | 634 | ||

Esrrb
Genomic Map