siRNA Details
| Gene Symbol: ZNF148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021964.2 | NP_068799.2 | 9 | 1868 | ||
| GenBank mRNA | AF039019.1 | - | 8 | 1593 | ||
| AF432210.1 | - | 9 | 1684 | |||
| AJ236885.1 | - | 9 | 1773 | |||
| AK314203.1 | - | 9 | 1803 | |||
| AL833616.1 | - | 9 | 1706 | |||
| BC050260.1 | - | 10 | 1944 | 4423572 | ||
| L04282.1 | - | 9 | 1770 | |||

ZNF148
Genomic Map