siRNA Details
| Gene Symbol: ZNF148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021964.2 | NP_068799.2 | 6 | 1040 | ||
| GenBank mRNA | AF039019.1 | - | 5 | 765 | ||
| AF432210.1 | - | 6 | 856 | |||
| AJ236885.1 | - | 6 | 945 | |||
| AK314203.1 | - | 6 | 975 | |||
| AL833616.1 | - | 6 | 877 | |||
| BC035591.1 | - | 6 | 951 | |||
| BC050260.1 | - | 7 | 1116 | 4423572 | ||
| BQ441085.1 | - | 3 | 294 | |||
| L04282.1 | - | 6 | 944 | |||

ZNF148
Genomic Map