siRNA Details
| Gene Symbol: LOC299282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182474.1 | NP_872280.1 | 10 | 1709 | ||
| GenBank mRNA | AY298742.1 | - | 10 | 1709 | ||
| AY325133.1 | - | 10 | 1709 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC299282| Gene Symbol: LOC299282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182474.1 | NP_872280.1 | 10 | 1709 | ||
| GenBank mRNA | AY298742.1 | - | 10 | 1709 | ||
| AY325133.1 | - | 10 | 1709 | |||