siRNA Details
| Gene Symbol: ZNF154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085384.2 | NP_001078853.1 | 3 | 845 | ||
| NR_110974.1 | 3 | 845 | ||||
| NR_110975.1 | 3 | 845 | ||||
| GenBank mRNA | AB095924.1 | - | 3 | 802 | ||
| AK092261.1 | - | 3 | 830 | |||
| BC152404.1 | - | 3 | 802 | 8862507 | ||
| BC152426.1 | - | 3 | 802 | 8862540 | ||
| BC152561.1 | - | 3 | 802 | 8860493 | ||
| DB162039.1 | - | 1 | 262 | |||

ZNF154
Genomic Map