siRNA Details
| Gene Symbol: Ndufb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106756.1 | NP_001100226.1 | 3 | 661 | ||
| GenBank mRNA | FQ211881.1 | - | 3 | 376 | ||
| FQ217020.1 | - | 3 | 377 | |||
| FQ217041.1 | - | 3 | 375 | |||
| FQ217852.1 | - | 3 | 381 | |||
| FQ223751.1 | - | 3 | 379 | |||
| FQ223793.1 | - | 3 | 407 | |||
| FQ224251.1 | - | 3 | 376 | |||
| FQ224299.1 | - | 3 | 377 | |||
| FQ224538.1 | - | 3 | 390 | |||

Ndufb11
Genomic Map