siRNA Details
| Gene Symbol: Apopt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037769.2 | NP_001032858.1 | 480 | |||
| GenBank mRNA | BC107661.1 | - | 475 | 7936717 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apopt1| Gene Symbol: Apopt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037769.2 | NP_001032858.1 | 480 | |||
| GenBank mRNA | BC107661.1 | - | 475 | 7936717 | ||