siRNA Details
Gene Symbol: ZNF157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003446.3 | NP_003437.2 | 4 | 1106 | ||
GenBank mRNA | BC075003.2 | - | 4 | 1078 | 30915612 | |
U28687.1 | - | 4 | 1123 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003446.3 | NP_003437.2 | 4 | 1106 | ||
GenBank mRNA | BC075003.2 | - | 4 | 1078 | 30915612 | |
U28687.1 | - | 4 | 1123 |