siRNA Details
| Gene Symbol: VEZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007146.2 | NP_009077.2 | 3 | 886 | ||
| XM_005257643.1 | XP_005257700.1 | 3 | 865 | |||
| XM_005257644.4 | XP_005257701.1 | 4 | 1021 | |||
| XM_017025017.1 | XP_016880506.1 | 4 | 1090 | |||
| XM_017025018.1 | XP_016880507.1 | 4 | 942 | |||
| GenBank mRNA | BX648862.1 | - | 2 | 185 | ||
| D28118.1 | - | 3 | 784 | |||

VEZF1
Genomic Map