siRNA Details
| Gene Symbol: Olr1078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207597.1 | NP_997480.1 | 1 | 771 | ||
| GenBank mRNA | M64376.1 | - | 1 | 771 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1078| Gene Symbol: Olr1078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207597.1 | NP_997480.1 | 1 | 771 | ||
| GenBank mRNA | M64376.1 | - | 1 | 771 | ||