siRNA Details
| Gene Symbol: Theg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013102.1 | NP_001013120.1 | 5 | 634 | ||
| XM_006240857.2 | XP_006240919.1 | 5 | 654 | |||
| XM_006240858.2 | XP_006240920.1 | 4 | 582 | |||
| GenBank mRNA | BC083921.1 | - | 5 | 634 | 7135133 | |

Theg
Genomic Map