siRNA Details
| Gene Symbol: Mex3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241041.2 | XP_006241103.2 | 1 | 639 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mex3d| Gene Symbol: Mex3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241041.2 | XP_006241103.2 | 1 | 639 | ||