siRNA Details
| Gene Symbol: Atp8b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_234937.8 | XP_234937.3 | 13 | 1286 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp8b3| Gene Symbol: Atp8b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_234937.8 | XP_234937.3 | 13 | 1286 | ||