siRNA Details
| Gene Symbol: Mknk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011985.1 | NP_001011985.1 | 10 | 854 | ||
| XM_006240883.2 | XP_006240945.1 | 10 | 943 | |||
| XM_006240884.1 | XP_006240946.1 | 9 | 831 | |||
| GenBank mRNA | BC085941.1 | - | 10 | 854 | 7124839 | |
| FQ211808.1 | - | 10 | 897 | |||

Mknk2
Genomic Map