siRNA Details
| Gene Symbol: Gadd45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008321.1 | NP_001008322.1 | 3, 4 | 606 | ||
| GenBank mRNA | BC085337.1 | - | 3, 4 | 606 | 7302492 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gadd45b| Gene Symbol: Gadd45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008321.1 | NP_001008322.1 | 3, 4 | 606 | ||
| GenBank mRNA | BC085337.1 | - | 3, 4 | 606 | 7302492 | |