siRNA Details
| Gene Symbol: Tle2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039013.1 | NP_001034102.1 | 11 | 887 | ||
| XM_006240889.2 | XP_006240951.1 | 11 | 1059 | |||
| XM_006240890.2 | XP_006240952.1 | 11 | 1536 | |||
| XM_006240892.2 | XP_006240954.1 | 11 | 972 | |||
| XM_006240893.2 | XP_006240955.1 | 11 | 1056 | |||
| XM_006240894.2 | XP_006240956.1 | 11 | 1059 | |||
| XM_006240895.2 | XP_006240957.1 | 11 | 1128 | |||
| GenBank mRNA | BC100652.1 | - | 11 | 887 | 7932424 | |

Tle2
Genomic Map