siRNA Details
| Gene Symbol: LOC100362054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758861.1 | XP_008757083.1 | 6 | 2296 | ||
| Gene Symbol: Zfp494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_234948.3 | XP_234948.1 | 3 | 1340 | ||

LOC100362054
Genomic Map