siRNA Details
| Gene Symbol: Vom2r57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099474.1 | NP_001092944.1 | 4 | 1379 | ||
| XM_008765199.1 | XP_008763421.1 | 5 | 1388 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom2r57| Gene Symbol: Vom2r57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099474.1 | NP_001092944.1 | 4 | 1379 | ||
| XM_008765199.1 | XP_008763421.1 | 5 | 1388 | |||
Set Membership: |
|