siRNA Details
| Gene Symbol: Bpifc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106773.1 | NP_001100243.1 | 3, 4 | 309 | ||
| XM_008765223.1 | XP_008763445.1 | 2, 3 | 468 | |||
| XM_008765224.1 | XP_008763446.1 | 2, 3 | 468 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bpifc| Gene Symbol: Bpifc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106773.1 | NP_001100243.1 | 3, 4 | 309 | ||
| XM_008765223.1 | XP_008763445.1 | 2, 3 | 468 | |||
| XM_008765224.1 | XP_008763446.1 | 2, 3 | 468 | |||