siRNA Details
| Gene Symbol: Bpifc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106773.1 | NP_001100243.1 | 13, 14 | 1332 | ||
| XM_008765223.1 | XP_008763445.1 | 12, 13 | 1491 | |||
| XM_008765224.1 | XP_008763446.1 | 12, 13 | 1485 | |||

Bpifc
Genomic Map