siRNA Details
| Gene Symbol: Cry1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198750.2 | NP_942045.2 | 9 | 1876 | ||
| XM_008765225.1 | XP_008763447.1 | 9 | 1900 | |||
| GenBank mRNA | AF545855.1 | - | 9 | 1440 | ||
| BC107677.1 | - | 9 | 1876 | 7939194 | ||

Cry1
Genomic Map