siRNA Details
| Gene Symbol: Ccdc53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106776.1 | NP_001100246.1 | 5 | 527 | ||
| XM_006241199.2 | XP_006241261.1 | 5 | 496 | |||
| GenBank mRNA | FQ221863.1 | - | 4 | 302 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc53| Gene Symbol: Ccdc53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106776.1 | NP_001100246.1 | 5 | 527 | ||
| XM_006241199.2 | XP_006241261.1 | 5 | 496 | |||
| GenBank mRNA | FQ221863.1 | - | 4 | 302 | ||