siRNA Details
| Gene Symbol: ZNF177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172651.1 | NP_001166122.1 | 6 | 1385 | ||
| NM_003451.2 | NP_003442.2 | 9 | 1200 | |||
| GenBank mRNA | AK302274.1 | - | 5 | 1290 | ||
| BC012012.1 | - | 5 | 791 | 4509206 | ||
| U37263.1 | - | 9 | 1200 | |||
| Gene Symbol: ZNF559-ZNF177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172650.2 | NP_001166121.1 | 12 | 1488 | ||
| NM_001202425.1 | NP_001189354.1 | 9 | 1780 | |||
| GenBank mRNA | U37251.1 | - | 9 | 1750 | ||

ZNF177
Genomic Map