siRNA Details
| Gene Symbol: Lta4h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030031.1 | NP_001025202.1 | 8 | 921 | ||
| XM_006241251.2 | XP_006241313.1 | 8 | 941 | |||
| GenBank mRNA | BC099819.1 | - | 8 | 921 | 7930840 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lta4h| Gene Symbol: Lta4h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030031.1 | NP_001025202.1 | 8 | 921 | ||
| XM_006241251.2 | XP_006241313.1 | 8 | 941 | |||
| GenBank mRNA | BC099819.1 | - | 8 | 921 | 7930840 | |