siRNA Details
| Gene Symbol: Amdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191781.1 | NP_001178710.1 | 5 | 700 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amdhd1| Gene Symbol: Amdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191781.1 | NP_001178710.1 | 5 | 700 | ||