siRNA Details
| Gene Symbol: Ndufa12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106781.2 | NP_001100251.2 | 1, 2 | 116 | ||
| GenBank mRNA | BF555697.1 | - | 1, 2 | 91 | ||
| FQ217295.1 | - | 1, 2 | 102 | |||
| FQ224477.1 | - | 1, 2 | 102 | |||
| Gene Symbol: LOC100910710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239763.2 | XP_006239825.1 | 1, 2 | 113 | ||

Ndufa12
Genomic Map