siRNA Details
| Gene Symbol: Mrpl42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106782.1 | NP_001100252.1 | 5 | 363 | ||
| XM_008765315.1 | XP_008763537.1 | 5 | 369 | |||
| GenBank mRNA | BC168747.1 | - | 5 | 351 | 6920919 | |
| Gene Symbol: LOC100361098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749244.3 | XP_003749292.1 | 2 | 274 | ||
| Gene Symbol: LOC103691922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769578.1 | XP_008767800.1 | 1 | 283 | ||

Mrpl42
Genomic Map