siRNA Details
| Gene Symbol: Nts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102381.1 | NP_001095851.1 | 2 | 188 | ||
| GenBank mRNA | FQ214250.1 | - | 2 | 261 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nts| Gene Symbol: Nts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102381.1 | NP_001095851.1 | 2 | 188 | ||
| GenBank mRNA | FQ214250.1 | - | 2 | 261 | ||