siRNA Details
Gene Symbol: Myf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106783.1 | NP_001100253.1 | 2, 3 | 814 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Myf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106783.1 | NP_001100253.1 | 2, 3 | 814 |