siRNA Details
| Gene Symbol: Glipr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011987.1 | NP_001011987.1 | 4, 5 | 750 | ||
| GenBank mRNA | BC089858.1 | - | 4, 5 | 750 | 7384037 | |
| FQ219892.1 | - | 4, 5 | 755 | |||
| FQ220342.1 | - | 4, 5 | 756 | |||
| FQ223414.1 | - | 4, 5 | 755 | |||
| FQ226490.1 | - | 4, 5 | 771 | |||
| FQ227021.1 | - | 4, 5 | 770 | |||
| FQ229493.1 | - | 3, 4 | 691 | |||
| FQ230706.1 | - | 4, 5 | 779 | |||

Glipr1
Genomic Map