siRNA Details
Gene Symbol: Glipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011987.1 | NP_001011987.1 | 1 | 323 | ||
GenBank mRNA | BC089858.1 | - | 1 | 323 | 7384037 | |
FQ219892.1 | - | 1 | 328 | |||
FQ220342.1 | - | 1 | 329 | |||
FQ223414.1 | - | 1 | 328 | |||
FQ226490.1 | - | 1 | 344 | |||
FQ227021.1 | - | 1 | 343 | |||
FQ229493.1 | - | 1 | 350 | |||
FQ229810.1 | - | 1 | 389 | |||
FQ230706.1 | - | 1 | 351 |
