siRNA Details
Gene Symbol: Glipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011987.1 | NP_001011987.1 | 1 | 209 | ||
GenBank mRNA | BC089858.1 | - | 1 | 209 | 7384037 | |
FQ219892.1 | - | 1 | 214 | |||
FQ220342.1 | - | 1 | 215 | |||
FQ223414.1 | - | 1 | 214 | |||
FQ226490.1 | - | 1 | 230 | |||
FQ227021.1 | - | 1 | 229 | |||
FQ229493.1 | - | 1 | 236 | |||
FQ229810.1 | - | 1 | 275 | |||
FQ230706.1 | - | 1 | 237 |
