siRNA Details
| Gene Symbol: Cct2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005905.1 | NP_001005905.1 | 7 | 543 | ||
| XM_006241371.2 | XP_006241433.1 | 7 | 592 | |||
| GenBank mRNA | BC083650.1 | - | 7 | 543 | 7135109 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cct2| Gene Symbol: Cct2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005905.1 | NP_001005905.1 | 7 | 543 | ||
| XM_006241371.2 | XP_006241433.1 | 7 | 592 | |||
| GenBank mRNA | BC083650.1 | - | 7 | 543 | 7135109 | |