siRNA Details
Gene Symbol: Yeats4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127527.2 | NP_001120999.1 | 2 | 177 | ||
GenBank mRNA | BC166753.1 | - | 2 | 177 | 5623659 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Yeats4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127527.2 | NP_001120999.1 | 2 | 177 | ||
GenBank mRNA | BC166753.1 | - | 2 | 177 | 5623659 |