siRNA Details
Gene Symbol: LOC100911956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764900.1 | XP_008763122.1 | 1 | 125 |
Gene Symbol: Llph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134464.2 | NP_001127936.1 | 1 | 91 | ||
XM_006241425.1 | XP_006241487.1 | 2 | 183 | |||
GenBank mRNA | BC158738.1 | - | 1 | 93 | 9099223 |
