siRNA Details
| Gene Symbol: Styxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241413.2 | XP_006241475.1 | 1 | 501 | ||
| Gene Symbol: Styx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271541.1 | NP_001258470.1 | 7, 8 | 599 | ||

Styxl2
Genomic Map