siRNA Details
| Gene Symbol: Lrig3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_216905.8 | XP_216905.5 | 8 | 1319 | ||
| GenBank mRNA | BC168689.1 | - | 8 | 1291 | 7932903 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrig3| Gene Symbol: Lrig3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_216905.8 | XP_216905.5 | 8 | 1319 | ||
| GenBank mRNA | BC168689.1 | - | 8 | 1291 | 7932903 | |