siRNA Details
| Gene Symbol: Tmem5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024759.1 | NP_001019930.1 | 6 | 1235 | ||
| XM_006241397.2 | XP_006241459.1 | 6 | 1300 | |||
| XM_008765391.1 | XP_008763613.1 | 5 | 987 | |||
| GenBank mRNA | BC097355.1 | - | 6 | 1235 | 7441970 | |

Tmem5
Genomic Map