siRNA Details
| Gene Symbol: Dctn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004239.1 | NP_001004239.1 | 11 | 987 | ||
| XM_006241448.1 | XP_006241510.1 | 12 | 1075 | |||
| XM_008765392.1 | XP_008763614.1 | 12 | 1077 | |||
| GenBank mRNA | BC079042.1 | - | 11 | 987 | 7114929 | |

Dctn2
Genomic Map