siRNA Details
| Gene Symbol: Shmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008322.1 | NP_001008323.1 | 8 | 928 | ||
| XM_006241453.2 | XP_006241515.1 | 8 | 954 | |||
| GenBank mRNA | BC085331.1 | - | 8 | 928 | 7106911 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shmt2| Gene Symbol: Shmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008322.1 | NP_001008323.1 | 8 | 928 | ||
| XM_006241453.2 | XP_006241515.1 | 8 | 954 | |||
| GenBank mRNA | BC085331.1 | - | 8 | 928 | 7106911 | |