siRNA Details
| Gene Symbol: Lrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130490.1 | NP_001123962.1 | 9 | 1742 | ||
| XM_008765393.1 | XP_008763615.1 | 9 | 1867 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrp1| Gene Symbol: Lrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130490.1 | NP_001123962.1 | 9 | 1742 | ||
| XM_008765393.1 | XP_008763615.1 | 9 | 1867 | |||