siRNA Details
| Gene Symbol: Ext1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130540.1 | NP_001124012.1 | 5 | 1504 | ||
| GenBank mRNA | BC105839.1 | - | 5 | 819 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ext1| Gene Symbol: Ext1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130540.1 | NP_001124012.1 | 5 | 1504 | ||
| GenBank mRNA | BC105839.1 | - | 5 | 819 | ||