siRNA Details
| Gene Symbol: Fam49b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126267.2 | NP_001119739.1 | 10, 11 | 967 | ||
| XM_006241632.2 | XP_006241694.1 | 8, 9 | 2079 | |||
| XM_006241633.1 | XP_006241695.1 | 9, 10 | 958 | |||
| XM_006241634.1 | XP_006241696.1 | 8, 9 | 817 | |||
| XM_006241635.2 | XP_006241697.1 | 9, 10 | 938 | |||
| XM_008765501.1 | XP_008763723.1 | 10, 11 | 1046 | |||
| GenBank mRNA | BC166469.1 | - | 10, 11 | 967 | 9036548 | |
| FQ212269.1 | - | 9, 10 | 963 | |||
| FQ229856.1 | - | 1, 2 | 76 | |||

Fam49b
Genomic Map