siRNA Details
| Gene Symbol: Fam49b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126267.2 | NP_001119739.1 | 7 | 582 | ||
| XM_006241632.2 | XP_006241694.1 | 5 | 1694 | |||
| XM_006241633.1 | XP_006241695.1 | 6 | 573 | |||
| XM_006241634.1 | XP_006241696.1 | 5 | 432 | |||
| XM_006241635.2 | XP_006241697.1 | 6 | 553 | |||
| XM_008765501.1 | XP_008763723.1 | 7 | 661 | |||
| GenBank mRNA | BC166469.1 | - | 7 | 582 | 9036548 | |

Fam49b
Genomic Map