siRNA Details
| Gene Symbol: Lrrc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025659.2 | NP_001020830.2 | 4 | 568 | ||
| GenBank mRNA | BC097394.1 | - | 4 | 568 | 7453303 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc6| Gene Symbol: Lrrc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025659.2 | NP_001020830.2 | 4 | 568 | ||
| GenBank mRNA | BC097394.1 | - | 4 | 568 | 7453303 | |